Learn More
Velp Scientifica™ TITROLINE 5000 Automatic Titrator
Product Code. 15349494 Shop All Velp Products
Change view
Click to view available options
Model:
Titroline Easy K
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
15349494 Titroline Easy K Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15349494 Supplier Velp Scientifica™ Supplier No. R30800194
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 18-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Velp Scientifica™ TITROLINE 5000 Automatic Titrator with compact new titrator for simple routine titrations.
  • Titrations to pH, mV - endpoint (2 EP)
  • Titrations to one equivalence point (pH und mV) with dynamic or linear titration addition
  • Manual titrations with the controller
  • Dosing tasks
  • Number of methods: 3
  • Salt content in foodstuffs (cheese, soy sauce, ketchup)
  • Total acidity in wine and beverages
  • Nitrogen according to Kjeldahl
  • Chloride in dringing water and other samples
  • Alkalinity (p + m-value)
  • FOS/TAC
Specifications
Display Type 3.5 in. color touch screen
Type Automatic Titrator
For Use With (Application) Combined distillation and titration, for simultaneous Kjeldahl nitrogen/protein determination
Material Technopolymer
Model Titroline Easy K
Depth (Metric) 416 mm