Learn More
TUTTNAUER Tiva 10 Glassware Washer with Manual Door
Product Code. 30058119 Shop All Tuttnauer Products
Change view
Click to view available options
Voltage:
380/400 V
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
30058119 380/400 V 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 30058119 Supplier TUTTNAUER Supplier No. TIVA101ML
  • ONLINE EXCLUSIVE PRICE
    22086.75€
    19557.60€ / 1 units
    Save 2529.15€
    11% Off
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Equipped with a manual double tempered glass door for laboratory glassware, featuring 4 independent washing levels.
  • Designed with 4 independent washing levels to efficiently clean a wide range of laboratory glassware.
  • The upper levels are removable, allowing easy accommodation of large laboratory materials.
  • Equipped with a powerful system that filters, electrically heats, and forces air through the wash chamber and inside hollow instruments for excellent drying results.
  • Features a durable double tempered glass manual door for enhanced safety and visibility during operation.
  • Includes a wide range of racks, nozzles, and accessories suitable to wash and dry various laboratory glassware types.
  • Large 265-liter chamber ensures optimal load capacity and cleaning efficiency.
Specifications
Display LCD
Material Stainless Steel
Product Type Glassware Washer with Drying System and Steam Condenser
Style Manual Door
Voltage 380/400 V