Learn More
Tizanidine hydrochloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30328570
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328570 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30328570 Supplier Dr. Ehrenstorfer Supplier No. DREA17587600AL100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tizanidine hydrochloride
CAS 64461-82-1
Analyte or Component Names Tizanidine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C9 H8 Cl N5 S . Cl H
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 5-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine monohydrochloride, 5-Chloro-4-(2-imidazolin-2-ylamino)-2,1,3-benzothiadiazole hydrochloride, AB 021, AN 021, DS 103-282, Sirdalud, Ternelin, Tizan, Tizanidine Hydrochloride, Tizpa, Zanaflex
InChI Formula InChI=1S/C9H8ClN5S.ClH/c10-5-1-2-6-8(15-16-14-6)7(5)13-9-11-3-4-12-9;/h1-2H,3-4H2,(H2,11,12,13);1H
SMILES Cl.Clc1ccc2nsnc2c1NC3=NCCN3
IUPAC Name 5-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine;hydrochloride
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 290.17
Quantity 1 mL
Formula Weight 288.9956 g/mol