missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TJAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17734-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a tight junction-associated protein. Incorporation of the encoded protein into tight junctions occurs at a late stage of formation of the junctions. The encoded protein localizes to the Golgi and may function in vesicle trafficking. Alternatively spliced transcript variants have been described. A related pseudogene exists on the X chromosome.Specifications
| TJAP1 | |
| Polyclonal | |
| Unconjugated | |
| TJAP1 | |
| PILT, Tight junction protein 4, TJAP1, TJP4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 93643 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5JTD0 | |
| TJAP1 | |
| TJAP1 Fusion Protein Ag12174 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction