missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TK1 Recombinant Rabbit Monoclonal Antibody (15A5)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550235
This item is not returnable.
View return policy
Description
TK1 Recombinant Monoclonal Antibody for IHC (P), ELISA
Thymidine Kinase (TK1) is a highly conserved phosphotransferase that is present in most living cells. Thymidine Kinase catalyzes the phosphorylation reaction: deoxythymidine + ATP = deoxythymidine 5'-phosphate + ADP; it is thus involved in the reaction chain to introduce deoxythymidine into the DNA. Thymidine kinase is required for the action of many antiviral drugs, such as azidothymidine (AZT), and is also used to select hybridoma cell lines in the production of monoclonal antibodies. Thymidine Kinase has many clinical applications as it is only present in anticipation of cell division.
Specifications
| TK1 | |
| Recombinant Monoclonal | |
| 2.65 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P04183 | |
| TK1 | |
| Recombinant Human TK1 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 15A5 | |
| Unconjugated | |
| TK1 | |
| D530002A18Rik; EC 2.7.1.21; KITH; thymidine kinase 1; thymidine kinase 1 soluble isoform; thymidine kinase 1, soluble; thymidine kinase, cytosolic; thymidine kinase-1; Tk; Tk1; Tk-1; Tk1a; Tk1b; TK2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7083 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction