Learn More
TLE1 Monoclonal Antibody (OTI3C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800320S
Description
TLE1 Monoclonal antibody specifically detects TLE1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| TLE1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q04724 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7088 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C9 | |
| Unconjugated | |
| TLE1 | |
| C230057C06Rik; E(Sp1) homolog; enhancer of split groucho; enhancer of split groucho-like protein 1; esg; ESG1; Estm14; grg1; Grg-1; Groucho-related protein 1; RP11-203D19.1; TLE 1; TLE1; tle1.S; tle3b; Tle4l; transducin like enhancer of split 1; transducin like enhancer of split 1 S homeolog; transducin-like enhancer of split 1 (E(sp1) homolog); transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 1, homolog of Drosophila E(spl); transducin-like enhancer of split 3 (E(sp1) homolog) b; transducin-like enhancer of split 4 (homolog of Drosophila E(spl)) - like; transducin-like enhancer protein 1; XELAEV_18001139mg; xgrg1 | |
| Human recombinant protein fragment corresponding to amino acids 180-460 of human TLE1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.