missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR3 Antibody (40C1285.6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 75 publications
244.00€ - 550.00€
Specifications
| Classification | Monoclonal |
|---|---|
| Form | Purified |
| Isotype | IgG1 κ |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18715643
|
Novus Biologicals
NBP2-24875SS |
244.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18700324
|
Novus Biologicals
NBP2-24875 |
0.1 mg |
550.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR3 Monoclonal specifically detects TLR3 in Human, Mouse, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), Bioactivity, Block/Neutralize.Specifications
| Monoclonal | |
| Purified | |
| IgG1 κ | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title