missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Antibody (76B357.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
309.00€ - 622.00€
Specifications
| Antigen | TLR4 |
|---|---|
| Clone | 76B357.1 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen 1:500, In vitro assay, Dot Blot, SDS-Page reported in scientific literature (PMID 33166339), Flow (Cell Surface), Flow (Intracellular), ELISA Capture (Matched Antibody Pair) 0.5 ug, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready, Knockout Validated, KnockDown Validated, Block/Neutralize, Dual RNAscope ISH-IHC |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoassay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), In vitro Assay, KnockDown, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18737893
|
Novus Biologicals
NB100-56566SS |
0.025 mg |
309.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18747783
|
Novus Biologicals
NB100-56566 |
0.1 mg |
622.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR4 Monoclonal specifically detects TLR4 in Human, Mouse, Rat, Porcine, Bovine, Mammal samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Dot Blot, SDS-Page, Flow (Cell Surface), Flow (Intracellular), ELISA Capture (Matched Antibody Pair), CyTOF-ready, Knockout Validated, Knockdown Validated, Block/Neutralize, Dual RNAscope ISH-IHC.Specifications
| TLR4 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoassay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), In vitro Assay, KnockDown, Neutralization | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
| O00206 | |
| 7099 | |
| This antibody was developed against a portion of amino acids 100-200 of human TLR4 (NP_612564). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 76B357.1 | |
| Western Blot 1-3 ug/ml, Flow Cytometry, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen 1:500, In vitro assay, Dot Blot, SDS-Page reported in scientific literature (PMID 33166339), Flow (Cell Surface), Flow (Intracellular), ELISA Capture (Matched Antibody Pair) 0.5 ug, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready, Knockout Validated, KnockDown Validated, Block/Neutralize, Dual RNAscope ISH-IHC | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine | |
| 76B357.1, ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, tlr4 76B357.1, TLR4 facs, TLR4 flow cytometry, TLR4 human, TLR4 ihc, TLR4 western blot, TOLL, toll-like receptor 4 | |
| TLR4 | |
| IgG2b κ | |
| Protein G purified | |
| The amino acid sequence used has 100% identity in human, chimp, baboon, Chinese hamster, and rat; 93% identity in bovine and pig; 85% identity in sheep and 78% identity in cat, horse and mouse. Human, Mouse, Rat. Ground squirrel reactivity reported by a customer review. Predicted cross-reactivity based on sequence identity: Chimp (100%), Baboon (100%), Chinese Hamster (100%), Bovine (93%), Pig (93%). Mammal reactivity reported in scientific literature (PMID: 25130694). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title