Learn More
Invitrogen™ TMA16 Recombinant Rabbit Monoclonal Antibody (HL3197)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556712
Product Code. 30275438
  • 500.00€ / 100µL
Description
TMA16 Recombinant Monoclonal Antibody for Western Blot
TMA16 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMA16
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Q96EY4
TMA16
Recombinant fragment of human FLJ11184.
100 μL
Primary
Human
Antibody
IgG
Western Blot
HL3197
Unconjugated
TMA16
1810029B16Rik; C4orf43; RGD1305222; TMA16; translation machinery associated 16 homolog; translation machinery associated 16 homolog (S. cerevisiae); translation machinery-associated protein 16; UPF0534 protein C4orf43; UPF0534 protein C4orf43 homolog
Rabbit
Protein A
RUO
55319
-20°C, Avoid Freeze/Thaw Cycles