Learn More
Invitrogen™ TMBIM1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5101938
Product Code. 16628414
  • 502.00€ / 100µL
Description
Antibody detects endogenous levels of total TMBIM1.
Negatively regulates aortic matrix metalloproteinase-9 (MMP9) production and may play a protective role in vascular remodeling.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMBIM1
Polyclonal
Unconjugated
TMBIM1
2310061B02Rik; AA960455; AU024746; C78899; Lfg3; mKIAA4161; MST100; MSTP100; PP1201; Protein lifeguard 3; Protein RECS1; Protein RECS1 homolog; PSEC0158; RECS1; Responsive to centrifugal force and shear stress gene 1 protein; Tmbib1; Tmbim1; transmembrane BAX inhibitor motif containing 1; transmembrane BAX inhibitor motif-containing protein 1
Rabbit
Affinity chromatography
RUO
316516, 64114, 69660
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8BJZ3, Q969X1
TMBIM1
A synthesized peptide derived from human TMBIM1(Accession Q969X1), corresponding to amino acid residues M251-T301.
100 μL
Primary
Human, Mouse, Rat