missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMBIM1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA567129

Product Code. 15929804

  • 566.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MSNPSAPPPYE DRNPLYPGPP Highest antigen sequence identity to the following orthologs - mouse 90%, rat 86%.

Negatively regulates aortic matrix metalloproteinase-9 (MMP9) production and may play a protective role in vascular remodeling.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TMBIM1
Polyclonal
Unconjugated
TMBIM1
2310061B02Rik; AA960455; AU024746; C78899; Lfg3; mKIAA4161; MST100; MSTP100; PP1201; Protein lifeguard 3; Protein RECS1; Protein RECS1 homolog; PSEC0158; RECS1; Responsive to centrifugal force and shear stress gene 1 protein; Tmbib1; Tmbim1; transmembrane BAX inhibitor motif containing 1; transmembrane BAX inhibitor motif-containing protein 1
Rabbit
Antigen affinity chromatography
RUO
64114
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q969X1
TMBIM1
Recombinant Human TMBIM1. Recombinant protein control fragment (Product #RP-107776).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.