missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMC1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00€ - 463.00€
Specifications
| Antigen | TMC1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667578
|
Novus Biologicals
NBP3-05609-100ul |
100 μg |
463.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683799
|
Novus Biologicals
NBP3-05609-20ul |
20 μg |
188.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMC1 Polyclonal antibody specifically detects TMC1 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| TMC1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 117531 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| DFNA36, DFNB11, DFNB7, transmembrane channel-like 1, transmembrane channel-like protein 1, Transmembrane cochlear-expressed protein 1, transmembrane, cochlear expressed, 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 600-700 of human TMC1 (NP_619636.2). INILRLHTSMYFQCWAVMCCNVPEARVFKASRSNNFYLGMLLLILFLSTMPVLYMIVSLPPSFDCGPFSGKNRMFEVIGETLEHDFPSWMAKILRQLSNPG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title