missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEFF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11928-1-AP-20UL
This item is not returnable.
View return policy
Description
TMEFF2 may be a survival factor for hippocampal and mesencephalic neurons. The shedded form up-regulates cancer cell proliferation, probably by promoting ERK1/2 phosphorylation.Specifications
| TMEFF2 | |
| Polyclonal | |
| Unconjugated | |
| TMEFF2 | |
| HPP1, TENB2, TMEFF2, Tomoregulin 2, TPEF, TR, TR 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23671, 363228, 56363 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9QYM9, Q9UIK5 | |
| Tmeff2 | |
| TMEFF2 Fusion Protein Ag2609 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction