missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM103 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98169UV
This item is not returnable.
View return policy
Description
TMEM103 Polyclonal antibody specifically detects TMEM103 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
TMEM103 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
54859 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
DyLight 350 | |
Angiotonin-transactivated protein 1, ATP1, C3orf75, chromosome 3 open reading frame 75, elongator acetyltransferase complex subunit 6, ELP6, FLJ20211, Protein TMEM103, TMEM103, transmembrane protein 103 | |
Produced in rabbits immunized with E. coli-derived Human TMEM103 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction