missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM108 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
113.00€ - 363.00€
Specifications
| Antigen | TMEM108 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 1:100-1:500 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615394
|
Novus Biologicals
NBP3-21447-100ul |
100 μg |
363.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650625
|
Novus Biologicals
NBP3-21447-25ul |
25 μg |
113.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM108 Polyclonal antibody specifically detects TMEM108 in Human samples. It is validated for ImmunofluorescenceSpecifications
| TMEM108 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| cancer/testis antigen 124, CT124, KIAA1690, MGC3040, transmembrane protein 108 | |
| Produced in rabbits immunized with E. coli-derived Human TMEM108 fragment | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.0 | |
| 66000 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title