missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM109 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18464C
530.40 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
TMEM109 Polyclonal antibody specifically detects TMEM109 in Human samples. It is validated for Western Blot
Specifications
TMEM109 | |
Polyclonal | |
Western Blot | |
transmembrane protein 109 | |
Recognizes region between residues Between Between 25 and 75.. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
79073 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction