Learn More
TMEM165 Mouse, anti-Human, Clone: 3B11C9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68584-1-IG-20UL
Description
The TMEM165 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TMEM165. This antibody recognizes human antigen. The TMEM165 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TMEM165 | |
| 3B11C9 | |
| TMEM165, TMPT27, TPARL, transmembrane protein 165, Transmembrane protein PT27, Transmembrane protein TPARL | |
| 20 μL | |
| 55858 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |