Learn More
TMEM165 Mouse, anti-Human, Clone: 3B11C9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68584-1-IG-20UL
 View more versions of this product
Product Code. 18036215
  • 145.00€ / 20µL
Description
The TMEM165 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TMEM165. This antibody recognizes human antigen. The TMEM165 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TMEM165
3B11C9
TMEM165, TMPT27, TPARL, transmembrane protein 165, Transmembrane protein PT27, Transmembrane protein TPARL
20 μL
55858
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa