missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM165 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20485-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of TM165 remains unknown.Specifications
| TMEM165 | |
| Polyclonal | |
| Unconjugated | |
| TMEM165 | |
| TMEM165, TMPT27, TPARL, transmembrane protein 165, Transmembrane protein PT27, Transmembrane protein TPARL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21982, 364137, 55858 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52875, Q4V899, Q9HC07 | |
| Tmem165 | |
| TMEM165 Fusion Protein Ag14362 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction