missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM16A/DOG1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12652-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can recognize isoform1 and isoform2 of human ANO1.
DOG-1, the novel marker, is expressed ubiquitously in gastrointestinal stromal tumors irrespective of kit or PDGFR alpha mutation status.Specifications
| TMEM16A/DOG1 | |
| Polyclonal | |
| Unconjugated | |
| ANO1 | |
| ANO1, Anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A, TMEM16A/DOG1, Transmembrane protein 16A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 309135, 55107 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5XXA6 | |
| Ano1 | |
| TMEM16A/DOG1 Fusion Protein Ag3320 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction