missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM214 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82033AF405
This item is not returnable.
View return policy
Description
TMEM214 Polyclonal antibody specifically detects TMEM214 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| TMEM214 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 54867 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 405 | |
| FLJ20254, FLJ39682, PP446, transmembrane protein 214 | |
| TMEM214 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human TMEM214. The immunogen is located within amino acids 70 - 120 of TMEM214. Amino Acid Squence: PAVEPKKPGNKKQPKKV | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction