missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM258 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
288.00€ - 589.00€
Specifications
| Antigen | C11orf10 |
|---|---|
| Concentration | 0.4mg/mL |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18499171
|
Novus Biologicals
NBP1-91711-25ul |
25 μL |
288.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18020717
|
Novus Biologicals
NBP1-91711 |
0.1 mL |
589.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM258 Polyclonal specifically detects TMEM258 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| C11orf10 | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| chromosome 11 open reading frame 10, protein NEF1 | |
| TMEM258 | |
| IgG | |
| Affinity Purified | |
| Specificity of human TMEM258 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.4mg/mL | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 746 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MELEAMSRYTSPVNPAVFPHLTVVL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title