missing translation for 'onlineSavingsMsg'
Learn More

TMEM266 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

415.00€ - 624.00€

Specifications

Antigen TMEM266
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18628907
View Documents
Novus Biologicals
NBP2-68685-25ul
25 μL
415.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18684538
View Documents
Novus Biologicals
NBP2-68685
100 μg
624.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TMEM266 Polyclonal antibody specifically detects TMEM266 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

TMEM266
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
chromosome 15 open reading frame 27, FLJ30304, transmembrane protein C15orf27
This antibody was developed against a recombinant protein corresponding to amino acids: YVLPVKLEMEMVIQQYEKAKVIQDEQLERLTQICQEQGFEIRQLRAHLAQQDLDLAAEREAALQAPHVLSQPRSRFKVLE
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Polyclonal
Purified
RUO
PBS (pH 7.2) and 40% Glycerol
123591
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.