Learn More
Invitrogen™ TMEM43 Recombinant Rabbit Monoclonal Antibody (24GB2325)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557797
 View more versions of this product
Product Code. 30275219
  • 198.00€ / 20µL
Description
TMEM43 Recombinant Monoclonal Antibody for Western Blot, Flow
This gene belongs to the TMEM43 family. Defects in this gene are the cause of familial arrhythmogenic right ventricular dysplasia type 5 (ARVD5), also known as arrhythmogenic right ventricular cardiomyopathy type 5 (ARVC5). Arrhythmogenic right ventricular dysplasia is an inherited disorder, often involving both ventricles, and is characterized by ventricular tachycardia, heart failure, sudden cardiac death, and fibrofatty replacement of cardiomyocytes. This gene contains a response element for PPAR gamma (an adipogenic transcription factor), which may explain the fibrofatty replacement of the myocardium, a characteristic pathological finding in ARVC.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMEM43
Recombinant Monoclonal
Unconjugated
TMEM43
1200015A22Rik; ARVC5; ARVD5; EDMD7; LUMA; Protein LUMA; Tmem43; Transmembrane protein 43; UNQ2564/PRO6244
Rabbit
Affinity chromatography
RUO
362401, 74122, 79188
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2325
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q5XIP9, Q9BTV4, Q9DBS1
TMEM43
A synthesized peptide derived from human TMEM43 (aa 100-135).