missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM52B Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98159C
This item is not returnable.
View return policy
Description
TMEM52B Polyclonal antibody specifically detects TMEM52B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TMEM52B | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 120939 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| C12orf59, chromosome 12 open reading frame 59, UNQ5927/PRO19821 | |
| Produced in rabbits immunized with E. coli-derived Human TMEM52B fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction