missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM87A Monoclonal Antibody (772807)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561891
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
| TMEM87A | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8NBN3 | |
| TMEM87A | |
| E. coli-derived recombinant human TMEM87A, Ser22-Asn157. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Western Blot | |
| 772807 | |
| Unconjugated | |
| TMEM87A | |
| PSEC0094; TMEM87A; Transmembrane protein 87A | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 25963 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction