missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMPRSS15 Monoclonal Antibody (01), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543526
This item is not returnable.
View return policy
Description
Specificity: Bovine Enterokinase/PRSS7
This gene encodes an enzyme that converts the pancreatic proenzyme trypsinogen to trypsin, which activates other proenzymes including chymotrypsinogen and procarboxypeptidases. The precursor protein is cleaved into two chains that form a heterodimer linked by a disulfide bond. This protein is a member of the trypsin family of peptidases. Mutations in this gene cause enterokinase deficiency, a malabsorption disorder characterized by diarrhea and failure to thrive. [provided by RefSeq, Jul 2008].
Specifications
| TMPRSS15 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P98072 | |
| TMPRSS15 | |
| Recombinant Bovine Enterokinase/PRSS7 Protein. | |
| 50 μL | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG1 |
| ELISA | |
| 01 | |
| Unconjugated | |
| TMPRSS15 | |
| enterokinase; enterokinase catalytic subunit; Enterokinase light chain; enteropeptidase; Enteropeptidase catalytic light chain; Enteropeptidase non-catalytic heavy chain; Entk; protease, serine, 7 (enterokinase); PRSS7; Serine protease 7; Tmprss15; Transmembrane protease serine 15; transmembrane protease, serine 15 | |
| Mouse | |
| Protein A | |
| RUO | |
| 282009 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction