Learn More
TMPRSS2 Antibody (CL12511) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18579C
Product Code. 30295683
  • 791.00€ / 0.10mL
Description
TMPRSS2 Monoclonal antibody specifically detects TMPRSS2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TMPRSS2
Monoclonal
DyLight 650
EC 3.4.21, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2
This antibody was developed using a recombinant protein derived from O15393, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Biology, Neuroscience, Prostate Cancer, Virology Bacteria and Parasites
7113
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL12511
50mM Sodium Borate
Mouse
Protein A purified