missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMPRSS9 Monoclonal Antibody (1042528)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561581
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a membrane-bound type II serine polyprotease that is cleaved to release three different proteases. Two of the proteases are active and can be inhibited by serine protease inhibitors, and one is thought to be catalytically inactive. This gene enhances the invasive capability of pancreatic cancer cells and may be involved in cancer progression.
Specifications
| TMPRSS9 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q7Z410 | |
| TMPRSS9 | |
| E. coli-derived recombinant human TMPRSS9, Glu190-Gln490. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 1042528 | |
| Unconjugated | |
| TMPRSS9 | |
| polyserase 1; polyserase-1; Polyserase-I; polyserine protease 1; polyserine protease-1; Serase-1; serase-1b; Serase-2; Serase-3; TMPRSS9; Transmembrane protease serine 9; transmembrane protease, serine 9; transmembrane serine protease 9 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 360200 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction