missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMSB10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550533
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total Thymosin beta 10 protein.
This protein plays an important role in the organization of the cytoskeleton. It binds to and sequesters actin monomers (G actin) and therefore inhibits actin polymerization. Experiment indicate that the expression decrease dramatically after birth; It expresses high levels in human fetal brain.Specifications
TMSB10 | |
Polyclonal | |
Unconjugated | |
TMSB10 | |
MIG12; migration-inducing gene 12; migration-inducing protein 12; prothymosin beta 10; Ptmb10; Tb10; Thyb10; thymosin; thymosin beta 10; thymosin beta-10; thymosin, beta 10; TMSB10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19240, 50665, 9168 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P63312, P63313, Q6ZWY8 | |
TMSB10 | |
Synthetic peptide corresponding to residues near the C terminal of human thymosin beta 10 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |