missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMTC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62592
This item is not returnable.
View return policy
Description
TMTC2 Polyclonal antibody specifically detects TMTC2 in Human samples. It is validated for Western Blot
Specifications
| TMTC2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| DKFZp762A217, transmembrane and tetratricopeptide repeat containing 2, transmembrane and TPR repeat-containing protein 2 | |
| Synthetic peptides corresponding to TMTC2(transmembrane and tetratricopeptide repeat containing 2) The peptide sequence was selected form the N terminal of TMTC2. Peptide sequence SNSDNPAADSDSLLTRTLTFFYLPTKNLWLLLCPDTLSFDWSMDAVPLLK. The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1.0 μg/mL | |
| Q8N394 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 160335 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction