missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF-alpha Antibody (6N1E7) - Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
177.00€ - 584.00€
Specifications
| Antigen | TNF-alpha |
|---|---|
| Clone | 6N1E7 |
| Concentration | 0.5mg/mL |
| Dilution | Western Blot 2-4ug/ml, Flow Cytometry 1ul/1 million cells, Blocking/Neutralizing 10ug/ml, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Blocking Assay, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18145262
|
Novus Biologicals
NBP2-27224SS |
0.025 mg |
177.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18136882
|
Novus Biologicals
NBP2-27224 |
0.1 mg |
584.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, Block/Neutralize.Specifications
| TNF-alpha | |
| 0.5mg/mL | |
| Western Blot, Flow Cytometry, Blocking Assay, CyTOF | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Asthma, Autoimmune Diseases, Autophagy, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
| P01375 | |
| 7124 | |
| Full-length recombinant human TNF alpha was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 6N1E7 | |
| Western Blot 2-4ug/ml, Flow Cytometry 1ul/1 million cells, Blocking/Neutralizing 10ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| TNF | |
| IgG1 κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title