missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNF RI/TNFRSF1A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
181.00€ - 441.00€
Specifications
| Antigen | TNF RI/TNFRSF1A |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 - 5 μg/ml, Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18207636
|
Novus Biologicals
NBP2-78785SS |
0.025 mg |
181.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18280138
|
Novus Biologicals
NBP2-78785 |
0.1 mg |
441.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNF RI/TNFRSF1A Polyclonal specifically detects TNF RI/TNFRSF1A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TNF RI/TNFRSF1A | |
| Western Blot 1 - 5 μg/ml, Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| CD120a, CD120a antigen, FPF, p55, p55-R, p60, sTNF RI, sTNF RI/TNFRSF1A, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor | |
| TNFRSF1A | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Death Receptor Signaling Pathway, Phospho Specific, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| 7132 | |
| Partial recombinant protein made to an N-terminal portion of the human TNFR1 protein (amino acids 2-211) [UniProt P19438] | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title