missing translation for 'onlineSavingsMsg'
Learn More

TNFAIP3 Mouse anti-Human, Rat, Clone: 3A11G6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66695-1-IG-150UL

 View more versions of this product

Product Code. 16838815

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene was identified as a gene whose expression is rapidly induced by the tumor necrosis factor (TNF). The protein encoded by this gene is a zinc finger protein, and has been shown to inhibit NF-kappa B activation as well as TNF-mediated apoptosis. Knockout studies of a similar gene in mice suggested that this gene is critical for limiting inflammation by terminating TNF-induced NF-kappa B responses.
Specifications

Specifications

TNFAIP3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P21580
TNFAIP3
TNFAIP3 Fusion Protein Ag18150
150 μL
Primary
Human, Rat
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Western Blot
3A11G6
Unconjugated
TNFAIP3
A20, OTUD7C, TNF alpha induced protein 3, TNFA1P2, TNFAIP3, Zinc finger protein A20
Mouse
Protein G
RUO
683206, 7128
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.