missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRH3/TNFRSF26 Antibody (509027), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5330H
This item is not returnable.
View return policy
Description
TNFRH3/TNFRSF26 Monoclonal antibody specifically detects TNFRH3/TNFRSF26 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TNFRH3/TNFRSF26 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 509027 | |
| Flow Cytometry, CyTOF-ready | |
| APO-3;DDR3;DR3;GEF720;LARD;PLEKHG5;TNFRSF12;TNFRSF25;TR3;TRAMP;Tumor necrosis factor receptor superfamily member 25;WSL-1;WSL-LR | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse TNFRH3/TNFRSF26, Ser20-Lys162, Accession # P83626 | |
| RUO | |
| 8718 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction