missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF13B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21520-1-AP-150UL
This item is not returnable.
View return policy
Description
Simultaneously four different laboratories identified a a new member of the tumor necrosis factor (TNF) family. This has been named as TALL-1 (1), THANK (TNF homologue that activates apoptosis, nuclear factor-kappaB, and c-Jun NH2-terminal kinase (2), BAFF (for B cell activating factor belonging to the TNF family) (3) and BlyS (B lymphocyte stimulator). Membrane-bound BAFF is processed and secreted through the action of a protease whose specificity matches that of the furin family of proprotein convertases. The receptor for BlyS/BAFF appears to be expressed predomitly on B cells and activated T cells. Recently two orphan receptors TACI (6) and BCMA (,8) have been shown to be receptors for BlyS/BAFF (6-9). TACI is expressed on B cells and signals through CAML, activating the transcription factors NF-AT, NF-kB, and AP-1. TACI is a type III transmembrane protein, as defined by the lack of a cleaved signal sequence and its N-terminal extracellular exposure. The cDNA codes for 293 amino acids with predicted molecular weight of approximately 32 kDa.Specifications
| TNFRSF13B | |
| Polyclonal | |
| Unconjugated | |
| TNFRSF13B | |
| CD267, CVID, TACI, TNFRSF13B, TNFRSF14B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23495 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14836 | |
| TNFRSF13B | |
| TNFRSF13B Fusion Protein Ag16097 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction