missing translation for 'onlineSavingsMsg'
Learn More

Tobramycin N’-Tetraethoxycarbonyl, TRC

Product Code. 30451050
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451050 50 mg 50mg
30456194 25 mg 25mg
30457183 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30451050 Supplier Toronto Research Chemicals Supplier No. TRCT52401550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tobramycin N’-Tetraethoxycarbonyl
Analyte or Component Names Tobramycin N’-Tetraethoxycarbonyl
Shipping Condition Room Temperature
Molecular Formula C30 H53 N5 O17
InChI Formula InChI=1S/C30H53N5O17/c1-5-45-27(41)32-13-9-14(33-28(42)46-6-2)24(52-26-21(39)19(35-30(44)48-8-4)20(38)18(12-36)50-26)22(40)23(13)51-25-15(34-29(43)47-7-3)10-16(37)17(11-31)49-25/h13-26,36-40H,5-12,31H2,1-4H3,(H,32,41)(H,33,42)(H,34,43)(H,35,44)/t13-,14+,15+,16-,17+,18+,19-,20+,21+,22-,23+,24-,25+,26+/m0/s1
Solution Type Neat
SMILES CCOC(=O)N[C@H]1C[C@@H](NC(=O)OCC)[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](NC(=O)OCC)[C@H]2O)[C@@H](O)[C@@H]1O[C@H]3O[C@H](CN)[C@@H](O)C[C@H]3NC(=O)OCC
IUPAC Name ethyl N-[(2R,3R,5S,6R)-6-(aminomethyl)-2-[(1R,2S,3S,4R,6S)-4,6-bis(ethoxycarbonylamino)-3-[(2S,3R,4S,5S,6R)-4-(ethoxycarbonylamino)-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-5-hydroxyoxan-3-yl]carbamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 755.765
Quantity 50 mg
Formula Weight 755.344
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.