missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Todd Hewitt Broth with Colistin and Nalidixic acid (LIM Broth)
Description
Todd-Hewitt Broth is ready to use medium originally described by Todd and Hewitt for the production of antigenic streptococcal haemolysin.
Specifications
Specifications
| Color | Sand Yellow, Transparent |
| Content And Storage | 2°C to 12°C |
| Format | Poured Tube |
| Manufacturing Origin | Germany |
| Product Type | Microbiology Media |
| Fill Weight | 4.5 to 5.5 g |
| Packaging Type | 50 Tubes/Box |
| Quantity | 50/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?