missing translation for 'onlineSavingsMsg'
Learn More

Tolnaftate, TRC

Product Code. 30457043
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457043 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457043 Supplier Toronto Research Chemicals Supplier No. TRCT5354001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tolnaftate
CAS 2398-96-1
Molecular Formula C19 H17 N O S
Quantity 1 g
Synonym Tolnaftate, O-Naphthalen-2-yl methyl(3-methylphenyl)carbamothioate
InChI Formula InChI=1S/C19H17NOS/c1-14-6-5-9-17(12-14)20(2)19(22)21-18-11-10-15-7-3-4-8-16(15)13-18/h3-13H,1-2H3
SMILES CN(C(=S)Oc1ccc2ccccc2c1)c3cccc(C)c3
IUPAC Name O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate
Molecular Weight (g/mol) 307.41
Formula Weight 307.1031
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.