missing translation for 'onlineSavingsMsg'
Learn More

Toluene-2,6-diisocyanate, 97%

Product Code. p-7046001
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11353346

Brand: Thermo Scientific Alfa Aesar L12745.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 91-08-7 | C9H6N2O2 | 174.159 g/mol

Toluene-2,6-diisocyanate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Toluene-2,6-diisocyanate is used as an organic chemical synthesis intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols, amines and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-08-7
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00002010
InChI Key RUELTTOHQODFPA-UHFFFAOYSA-N
Synonym 2,6-diisocyanatotoluene, toluene-2,6-diisocyanate, 2,6-toluene diisocyanate, 2-methyl-m-phenylene diisocyanate, 2,6-tdi, m-tolylene diisocyanate, meta-tolylene diisocyanate, toluene 2,6-diisocyanate, benzene, 1,3-diisocyanato-2-methyl, 2,6-diisocyanato-1-methylbenzene
PubChem CID 7040
ChEBI CHEBI:53557
IUPAC Name 1,3-diisocyanato-2-methylbenzene
SMILES CC1=C(C=CC=C1N=C=O)N=C=O

Specifications

Melting Point 13°C
Density 1.226
Boiling Point 129°C to 133°C (18 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.5708
Quantity 1 g
UN Number UN2078
Beilstein 2211546
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 174.16
Percent Purity 97%
Chemical Name or Material Toluene-2,6-diisocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.