missing translation for 'onlineSavingsMsg'
Learn More

Toluidine Blue O, TRC

Product Code. 30456000
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30456000 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30456000

Brand: Toronto Research Chemicals TRCT53628350G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Toluidine Blue
CAS 92-31-9
Molecular Formula C15 H16 N3 S . Cl
Quantity 50 g
Synonym Phenothiazin-5-ium','3-amino-7-(dimethylamino)-2-methyl-','chloride (1:1), Phenothiazin-5-ium','3-amino-7-(dimethylamino)-2-methyl-','chloride (8CI,9CI), 3-Amino-7-(dimethylamino)-2-methylphenazathionium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazi
InChI Formula InChI=1S/C15H16N3S.ClH/c1-9-6-13-15(8-11(9)16)19-14-7-10(18(2)3)4-5-12(14)17-13;/h4-8H,16H2,1-3H3;1H/q+1;/p-1
SMILES [Cl-].CN(C)c1ccc2nc3cc(C)c(N)cc3[s+]c2c1
IUPAC Name 7-N,7-N,2-trimethylphenothiazin-5-ium-3,7-diamine;chloride
Molecular Weight (g/mol) 305.826
Formula Weight 305.075
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.