missing translation for 'onlineSavingsMsg'
Learn More

TOMM22 Mouse anti-Human, Clone: 1C2B11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66562-1-IG-20UL

 View more versions of this product

Product Code. 16828555

  • 145.00€ / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

66562-1-Ig recognizes a22 kDa protein that is larger than the calculated MW of 15 kDa.

Tom22 has a negatively charged N-terminal region exposed to the cytosol, a putative transmembrane region, and a C-terminal intermembrane space region with little negative charge. The following functions can be assigned to the three domains of Tom22: the cytosolic N-terminal domain plays a dual role, specifically important for presequence binding; the intermembrane space domain provides a trans binding site for presequences, and the single membrane anchor of Tom22 is crucial for the integrity of the GIP (general import gene) complex. The TOM complex of mammalian mitochondria resembles the fungal Tom complex, but is distinct from the plant TOM system. Thus, while unique components of the mammalian mitochondrial import system have been identified (e. g. TOM34 and metaxin), Tom22, and Tom37 have not been identified in plant mitochondria.
Specifications

Specifications

TOMM22
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NS69
TOMM22
TOMM22 Fusion Protein Ag1797
20 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
1C2B11
Unconjugated
TOMM22
1C9 2, hTom22, MST065, MSTP065, TOM22, TOMM22
Mouse
Protein A
RUO
56993
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.