Learn More
Tork Tork Mini Centrefeed Dispenser White and Turquoise M1
Product Code. 32209941 Shop All Tork Products
Change view
Click to view available options
Holds:
Wipes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
32209941 Wipes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32209941 Supplier Tork Supplier No. 658000
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Easy-to-clean, robust design aimed to improve overall hygiene levels
  • Suit tough environments and tight spaces with robust dispenser and small footprint
  • Save staff time with simple to use one-handed operation
  • Minimise cross-contamination with free flow dispensing allowing users to take as much as needed
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mini Centrefeed Dispenser
Color White, Blue
Length (English) 7.047 in.
Length (Metric) 179 mm
Width (English) 8.031 in.
Width (Metric) 204 mm
Height (English) 13.6 in.
Height (Metric) 347 mm
Holds Wipes
Material ABS
Mounting Wall Mount
No. of Compartments 1
Product Style Hinged Lid with Round Opening
Model Centerfeed