missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Torsin 2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68915
This item is not returnable.
View return policy
Description
Torsin 2A Polyclonal antibody specifically detects Torsin 2A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Torsin 2A | |
| Polyclonal | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| FLJ14771, MGC99558, pro salusin, prosalusin, TORP1torsin-2A, Torsin family 2 member A, torsin family 2, member A, Torsin-2A, Torsin-related protein 1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AIFIFISNTGGEQINQVALEAWRSRRDREEILLQELEPVIS | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 27433 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction