Learn More
Ansell™ TouchNTuff™ 69-210 Series Natural Rubber Latex Gloves
Product Code. 15386957 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15386957 Medium Pack of 100
15376957 Small Pack of 100
15306967 Large Pack of 100
15316967 - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15386957 Supplier Ansell™ Supplier No. 69210080
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Powdered for easier donning and moisture absorption. Ansell TouchNTuff™ 69-210 Series Natural Rubber Latex Gloves offer superior tactile sensitivity for delicate tasks
  • Combined safety and comfort for delicate handling.
  • Ambidextrous, same glove fits both hands
  • Convenient and economical - stocks can be kept to a minimum
  • Flexible and comfortable to wear - cooler than vinyl, with superior finger flexibility and grip
  • Available in both powdered (69-210) and powder-free (69-318) versions
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420:2003 + A1:2009 and EN 374:2003
Color Natural
Latex Free No
Material Rubber Latex
Powdered Powdered
Product Type Natural Rubber Latex Gloves
Size Medium
Sterility Non-sterile
Thickness (Metric) 0.10 mm
Packaging Type 100 gloves per box, 10 boxes per carton/case
Quantity 100/box
Series 69-210
Length (Metric) Cuff Standard