Learn More
Ansell TouchNTuff™ 93-700 Series Green Nitrile Gloves
BIOPHARMA
Product Code. 13429028 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
200 pairs
Product Code. Size unitSize
13429028 10 200 pairs
13419028 7 200 pairs
13459028 8 200 pairs
13449028 9 200 pairs
4 options
Product Code. 13429028
Brand: Ansell 93700100
Only null left
Add to basket
Add to basket
Delivers the highest protection for a wide range of chemicals than any other single use nitrile glove. Manufactured from nitrile, these gloves offer three times the puncture resistance of standard latex gloves.
  • Provides the wearer with a high level of comfort and increased protection in the forearm with longer cuff.
  • Free of natural rubber latex proteins, the glove may be worn with confidence by latex sensitive (Type I) wearers.
  • 93-700: Each glove is post cleaned, single pair packed and sterilized.
TRUSTED_SUSTAINABILITY
Specifications
Cleanroom Class Class 100 (ISO 5)
Accelerator Free No
AQL 1.5
Color Green
Disposable Single-use
Exterior Finish Fully Textured
Hand Specific
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Controlled Environment Gloves
Size 10
Sterility Sterile
Thickness (Metric) 0.135 mm
For Use With (Application) For blending of liquids, filtration processes, vaccines injectables operations etc.
Model 9370010
Quantity 200/Pk.
Resistant To Splash and high puncture resistance
Series 93-700
Length (English) 12
Length (Metric) 300