missing translation for 'onlineSavingsMsg'
Learn More

Ansell TouchNTuff™ 93-700 Series Green Nitrile Gloves

Product Code. 16398265 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
Unit Size:
Pack of 200
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16398265 9 Pack of 200
16388265 7 Pack of 200
16308275 8 Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16398265 Supplier Ansell Supplier No. 93700090

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Delivers the highest protection for a wide range of chemicals than any other single use nitrile glove. Manufactured from nitrile, these gloves offer three times the puncture resistance of standard latex gloves.

  • Provides the wearer with a high level of comfort and increased protection in the forearm with longer cuff.
  • Free of natural rubber latex proteins, the glove may be worn with confidence by latex sensitive (Type I) wearers.
  • 93-700: Each glove is post cleaned, single pair packed and sterilized.
TRUSTED_SUSTAINABILITY

Specifications

Cleanroom Class Class 100 (ISO 5)
Accelerator Free No
AQL 1.5
Color Green
Disposable Single-use
Exterior Finish Fully Textured
Hand Specific
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Controlled Environment Gloves
Size 9
Sterility Sterile
Thickness (Metric) 0.135 mm
For Use With (Application) For blending of liquids, filtration processes, vaccines injectables operations etc.
Model 937009
Quantity 200/Pk.
Resistant To Splash and high puncture resistance
Series 93-700
Length (English) 12
Length (Metric) 300
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.