Learn More
Trajan™ Autosampler Syringes for Agilent GCs
Product Code. 12937556
Change view
Click to view available options
Volume (Metric):
10 μL
50 μL
Quantity:
1/Pk.
25/Pk.
6/Pk.
Unit Size:
Each
Pack of 25
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Quantity unitSize
12937556 50 μL 1/Pk. Each
10654643 10 μL 25/Pk. Pack of 25
10545603 10 μL 6/Pk. Pack of 6
12706897 10 μL 6/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12937556 Supplier Trajan™ Supplier No. 004668
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
Accurate and reproducible injections. Cone-tipped dual gauge needle. Available with or without Gas Tight plunger.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Agilent instruments
Specifications
Material Borosilicate Glass (Barrel), PTFE (Plunger)
Needle Gauge 23 to 26 G
Diameter (Metric) Inner Needle 0.11 mm
Diameter (Metric) Outer 6.5 mm
Diameter (Metric) Outer Needle 0.63/0.47 mm
Product Type Gas-tight GC Autosampler Syringe
Model 50F-HP-GT-0.63/47
Quantity 1/Pk.
Volume (Metric) 50 μL
Length (Metric) Needle 42 mm
Connection Type Fixed