missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPII Mouse anti-Human, Clone: 5H6F11, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | TPPII |
|---|---|
| Clone | 5H6F11 |
| Concentration | 1 mg/mL |
| Applications | Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16877515
|
Proteintech
66017-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16867515
|
Proteintech
66017-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.Specifications
| TPPII | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| TPP 2, TPP2, TPPII, Tripeptidyl aminopeptidase, Tripeptidyl peptidase 2, tripeptidyl peptidase II | |
| TPP2 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 5H6F11 | |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P29144 | |
| 7174 | |
| TPPII Fusion Protein Ag5296 | |
| Primary | |
| -20°C | |
| TPP2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title