missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TR 14035

Description
TR 14035 is a potent integrin α4β7 and α4β1 antagonist (IC50 values are 7 and 87 nM, respectively). Inhibits binding of HEK 293 cells to osteopontin forms. Orally bioavailable.
Specifications
Specifications
| Quantity | 50 mg |
| Molecular Formula | C24H21Cl2NO5 |
| Formula Weight | 474.33 |
| Product Type | Antagonist |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?