Learn More
TRA-1-60 (Podocalyxin) Mouse, anti-Human, PE, Clone: TRA-1-60R, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60064PE
Description
- The TRA‐1‐60R antibody reacts with TRA-1-60 (Podocalyxin), a > 200 kDa pluripotent stem cell-specific protein expressed on the surface of undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells, embryonal carcinoma (EC) cells and embryonic germ (EG) cells, as well as rhesus monkey ES cell lines
- The epitope contains sialic acid and is associated with a large-molecular-mass transmembrane protein named podocalyxin
- Though sialylated, the epitope recognized by the TRA‐1‐60R antibody is resistant to treatment with neuraminidase
- This antibody clone has been verified for labeling human ES and iPS cells grown in TeSR™-E8™ (Catalog #05940), mTeSR™1 (Catalog #85850), and TeSR™2 (Catalog #05860)
Specifications
| TRA-1-60 (Podocalyxin) | |
| Monoclonal | |
| PE | |
| 100 Tests | |
| Rhesus Monkey, Human, Rabbit | |
| Pluripotent Stem Cells |
| Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| TRA-1-60R | |
| Mouse | |
| Primary | |
| IgM κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.